Documenti fondamentali
LIDF34638-EP
Lid for Eppendorf® F-34-6-38 Rotor
About This Item
Codice UNSPSC:
41103913
Produttore/marchio commerciale
Eppendorf® 022662961
tecniche
centrifugation: suitable
Compatibilità
for use with 5430 R
for use with 5430
Categorie correlate
Note legali
Eppendorf is a registered trademark of Eppendorf AG