Documenti fondamentali
L5789
Sigma-Aldrich
Laser Tough-Spots®
diam. 3/8 in., red
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Materiali
polyolefin
red
Confezionamento
pack of 3840 ea (192 labels per sheet)
Produttore/marchio commerciale
Diversified Biotech SPOT-1400
Diametro
3/8 in.
Descrizione generale
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Note legali
Tough-Spots is a registered trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Tough-Spots® size 3/8 in., blue
Z740256
Tough-Spots®
Tough-Spots® size 3/8 in., white
Z740255
Tough-Spots®
Tough-Spots® 5 across, diam. 3/8 in., assorted colors, roll of 5000
Z743601
Tough-Spots®
Tough-Spots® diam. 1/2 in., lavender, roll of 1000
Z743592
Tough-Spots®
Tough-Spots® 5 across, diam. 1/2 in., assorted colors, roll of 5000
Z743588
Tough-Spots®
Laser Tough-Spots® diam. 3/8 in., assorted colors, pack of 3840 (192 labels per sheet)
Z743584
Laser Tough-Spots®
Tough-Spots® size 3/8 in., orange
Z740258
Tough-Spots®
Tough-Spots® size diam. 1/2 in, white, pkg of 1000 roll
Z688371
Tough-Spots®
Tough-Spots® size 1/2 in., white
Z740261
Tough-Spots®