Documenti fondamentali
GE18-1153-08
Tricorn Glass Tube
joint: 5/200
About This Item
Codice UNSPSC:
41115709
NACRES:
NB.43
Giunto
joint: 5/200
Produttore/marchio commerciale
Cytiva 18-1153-08
18-1153-13
Descrizione generale
Tricorn columns incorporate modern design and high performance to increase the user friendliness and reliability of the chromatography column. Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures.
  • High performance glass columns designed for reliable high resolution, reproducibility and durability
  • New longer adaptor for increased adjustment of bed height
  • Modern, integrated design – simple to assemble, pack, operate, and maintain
  • Broad range of column dimensions – suitable for many different purification needs
  • Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
  • Locking ring – prevents accidental compression of media bed
  • Separate packing connector – allows simple column packing for a broad range of media
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)