Documenti fondamentali
GBL505118
Grace Bio-Labs ONCYTE® NOVA nitrocellulose film slides
8- 6 mm x 20 mm NC pad per slide, plain glass
Sinonimo/i:
nitrocellulose slides, protein microarrays
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.15
Materiali
nitrocellulose (on glass)
Confezionamento
pack of 20 ea
Produttore/marchio commerciale
Grace Bio-Labs 505118
Dimens. esterne
25 mm × 75 mm
Tipo di binding
medium binding surface
Descrizione generale
Grace Bio-Labs ONCYTE® NOVA nitrocellulose coated substrates are designed for SBS compliant multiwell plates and glass substrates for high throughput automated processing of microarrays.
Note legali
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
GBL505118-20EA4061832399102