Key Documents
G1151
Sigma-Aldrich
Glass plates for vertical standard gel
16.5 cm × 28 cm
About This Item
Codice UNSPSC:
41105312
NACRES:
NB.51
product name
Glass plates for vertical standard gel, 16.5 cm wide, W × H 16.5 cm × 28 cm
Largh. × alt.
16.5 cm × 28 cm
Confezionamento
Set includes one rectangular plate with rounded bottom corners and one notched plate.