EPF571240035
Eppendorf® CryoCube® F570h
capacity 570 L, classic interface, cooling, 5, AC/DC input 208 V, 60 Hz, US 3-pin plug
Sinonimo/i:
CryoCube® F570h, CryoCube® F570h-ULT Freezer, Eppendorf® CryoCube® F570h-ULT Freezer
About This Item
Codice UNSPSC:
41103034
Prodotti consigliati
Caratteristiche
cooling 5
classic interface
Capacità
570 L
Descrizione generale
The Eppendorf CryoCube F740 series is one of the newest additions to the Eppendorf ultra-low temperature freezer family (also called ultra-low temperature freezer, -80C). The CryoCube F740 series of ultra-low temperature freezers combines the traditionally high quality of our previous ultra-low temperature freezers with a higher capacity to safely store more of your samples while optimizing energy consumption. By incorporating the PhysioCare Concept into the development, the CryoCube F740 is lighter and more comfortable to use, supports ergonomics in the workplace and enables an optimal workflow in the laboratory.
Note legali
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
EPF571240035-1EA | 4065268074121 |