EP951020539
Eppendorf® twin.tec PCR Plates 384
Optimized rim for barcodes, PCR clean, colorless, pack of 300 ea
About This Item
Codice UNSPSC:
41106300
Materiali
colorless
conical bottom
polypropylene wells
Sterilità
non-sterile
Caratteristiche
PCR clean
lid: no
skirt
Confezionamento
pack of 300 ea
Produttore/marchio commerciale
Eppendorf® 951020539
Misura
384 well
Volume pozzetto
45 μL
Compatibilità
suitable for PCR
Categorie correlate
Descrizione generale
- Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, colorless, 300 plates
- Optimized rim for barcodes and automated work processes
- Stackable
- Within SBS footprint recommendations
- Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
- 40 µL max. well volume
- Available with barcode (upon request), for skirted and semi-skirted only
Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, colorless, 300 plates
- Optimized rim for barcodes and automated work processes
- Stackable
- Within SBS footprint recommendations
- Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
- 40 µL max. well volume
Caratteristiche e vantaggi
- Optimized rim for barcodes and automated work processes
- Stackable
- Within SBS footprint recommendations
- Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
- 40 µL max. well volume
- Available with barcode (upon request), for skirted and semi-skirted only
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)