Documenti fondamentali
EP6731011815
Eppendorf® CellXpert® C170i CO2 Incubator
new
Copper interior, Oxygen control, water/humidity monitoring, inner door non-segmented, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
Sinonimo/i:
Heaters
About This Item
Codice UNSPSC:
41104405
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 6731011815
Descrizione generale
CellXpert® C170i, non-segmented inner door, handle right side, 1 – 20 % oxygen control, water and humidity monitoring package, copper package, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Caratteristiche e vantaggi
  • 1 – 20 % oxygen control
  • water and humidity monitoring package
  • handle right side
  • non-segmented inner door
  • 180° HTD (standard with C170i)
  • copper package: chamber, water tray and four shelves
  • 100 – 127 V, 50 – 60 Hz
Note legali
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)