Key Documents
EP5895155006
Rotor for Eppendorf® 5910 R Centrifuge
FA-30x2, incl. aerosol-tight QuickLock Rotor for Eppendorf(R) lid, for 30 x 2 mL tubes
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
Caratteristiche
to hold 30 x 1.5/2.0 mL tubes
FA-30x2, incl. aerosol-tight QuickLock Rotor for Eppendorf(R) lid, for 30 x 2 mL tubes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5895155006
Parametri
13,700 max. RCF (20,984 xg)
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5910R
Descrizione generale
Rotor for Eppendorf(R)5910R Centrifuge
Note legali
Eppendorf is a registered trademark of Eppendorf AG