Documenti fondamentali
EP5825706005
Eppendorf® Rotor Accessories
CombiSlide adapter, capacity, 12 slides
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.82
Caratteristiche
capacity 12 slides
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Eppendorf® 5825706005 (EU)
tecniche
centrifugation: suitable
Compatibilità
for use with MTP bucket
Note legali
Eppendorf is a registered trademark of Eppendorf AG