Documenti fondamentali
EP5810724007
Eppendorf® Aerosol-tight lid
for 500 mL rectangular bucket, pack of 2 ea
About This Item
Codice UNSPSC:
41103913
NACRES:
NB.82
Caratteristiche
for 500 mL rectangular bucket
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Eppendorf® 5810724007
tecniche
centrifugation: suitable
Compatibilità
for use with 5810 R
for use with 5810
Note legali
Eppendorf is a registered trademark of Eppendorf AG