Documenti fondamentali
EP5392070020
Eppendorf(R) Plate Adapter HeatSealer S200
for 96 and 384 well plates, low profile
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.15
Caratteristiche
for 96 and 384 well plates
low profile
Produttore/marchio commerciale
Eppendorf® 5392070020 (Global)
Categorie correlate
Caratteristiche e vantaggi
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)