EP4987000371
Eppendorf® Multipette® E3/E3x/M4
E3 bundle single-channel, with charging cable, charger stand, and Combitips advanced assortment pack (1 Combitip of each size), 1 ul - 50 ml, AC/DC input 100 V / 240 V, 50-60 Hz (EU)
Sinonimo/i:
Adjustable volume pipette
About This Item
Prodotti consigliati
Caratteristiche
CE compliant
E3 bundle single-channel
with charging cable, charger stand, and Combitips advanced assortment pack (1 Combitip of each size)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 4987000371 (EU)
Campo di misura
1 ul - 50 ml
Volume
1 μL × 50 mL
Batteria
rechargeable
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)