Documenti fondamentali
EP0030128818
Eppendorf® Twin tec PCR plates, 384 wells
yellow plate, conical bottom black wells, pkg of 25 ea
About This Item
Codice UNSPSC:
41106300
NACRES:
NB.15
Materiali
conical bottom black wells
yellow plate
Caratteristiche
skirt (Conical bottom)
Confezionamento
pkg of 25 ea
Produttore/marchio commerciale
Eppendorf® 0030128818
tecniche
PCR: suitable
Misura
384 wells
Descrizione generale
“Eppendorf” Twin tec PCR plates with 384 wells are large scale PCR plates used in high throughput PCR workflows.
Applicazioni
Eppendorf “twin.tec” PCR Plates may be used in a wide variety of applications such as:
  • Standard PCR
  • Real time PCR
  • High throughput PCR
Caratteristiche e vantaggi
  • [TM=“Eppendorf”] Twin tec PCR plates have a unique one - piece design consisting of polypropylene wells and a rigid polycarbonate framework.
  • Twin tec PCR plates have thin-walled polypropylene wells that ensure optimum heat transfer to the sample.
  • The exceptionally solid polycarbonate framework is torque resistant.
  • [TM=“Eppendorf”] Twin tec PCR plates show an improved well – well tolerance.
  • The presence of raised well rims ensures effective sealing.
  • [TM=“Eppendorf”] Twin tec PCR plates have been batch tested and certified DNase, RNase and PCR inhibitor free.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)