Key Documents
EP0030124839
Eppendorf® cap strips for PCR tubes
domed, 10 x 12 strips, pkg of 120 ea
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
domed
Caratteristiche
autoclavable 120 °C
domed
Confezionamento
pkg of 120 ea
Produttore/marchio commerciale
Eppendorf® 0030124839
Note legali
Eppendorf is a registered trademark of Eppendorf AG