Documenti fondamentali
EP0030081064
Eppendorf® BioBased ep Dualfilter T.I.P.S.
new
greener alternative
volume 2—100 μL, sterile, PCR clean, L -53 mm, pkg of 960 ea (10 trays x 96 tips)
About This Item
Codice UNSPSC:
41121600
Sterilità
sterile
Caratteristiche
DNase free
PCR clean
RNase free
graduations
Confezionamento
pkg of 960 ea (10 trays x 96 tips)
Produttore/marchio commerciale
Eppendorf® 0030081064
Caratteristiche più verdi
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
tecniche
DNA purification: suitable
PCR: suitable
Lunghezza
-53 mm
Volume
2—100 μL
Compatibilità
suitable for PCR
suitable for microbiology
suitable for molecular biology
suitable for nucleic acid purification
Categoria alternativa più verde
Descrizione generale
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product aligns with ″Use of Renewable Feedstocks″. Click here for more information.
Applicazioni
Molecular Biology, DNA purification, PCR, Microbiology, Cell Biology, Medical Research, Agriculture Research, Environmental Monitoring, Forensic Analysis
Caratteristiche e vantaggi
  • Improve your laboratory carbon footprint and switch to epT.I.P.S. BioBased in the new Eppendorf Reload variants. These pipette tips are produced from second-generation biobased polymer with 90% of the feedstock attributed to renewable resources (i.e. waste and residues from vegetable oilrefining or waste cooking oil), avoiding the use of fossil fuel-based petroleum.
  • Reduce your laboratory waste volume – use Reloads instead of disposable Racks.
  • Eppendorf production facility is ISCC PLUS(International Sustainability & Carbon Certification) certified, confirmingthe raw materials used at the facility are “bio-circular”. Enables traceabilityalong the entire supply chain.Pipette tips with ACT (Accountability, Consistency,and Transparency) Label - an independent environmental impact factor assessmentthrough non-profit My Green.
  • Material that is both quality and purity certified.
  • Get excellent sample integrity as no slip agents, plasticizers and biocides are used Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website.
  • General quality certificates in compliance with standards and general Eppendorf quality items.
  • Available in volumes from 10 µL to 1,250 µL Long in Reloads for use with epT.I.P.S. Box 2.0.
  • Available as epT.I.P.S. BioBased Biopur, ep Dualfilter T.I.P.S. BioBased PCR clean and sterile, ep Dualfilter T.I.P.S. SealMax BioBased Biopur
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)