CLS6383A100
Pyrex® cylindrical separatory funnel, PTFE stopcock
with standard taper joints, graduated, joint: ST/NS 24/40
Sinonimo/i:
Corning® 6383
About This Item
Codice UNSPSC:
41121809
Prodotti consigliati
Giunto
joint: ST/NS24/40
Caratteristiche
graduated
with standard taper joints
Produttore/marchio commerciale
Corning 6383A-100
Categorie correlate
Descrizione generale
Graduated, with ST Pyrex stopper and microfinish PTFE stopcock.
CLS6383A100 - does not include a stopper
CLS6383A100 - does not include a stopper
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)