Documenti fondamentali
CLS432129
Corning® Ice Bucket with lid, round
2.5 L, blue
Sinonimo/i:
storage container
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.12
Confezionamento
pkg of 1
Produttore/marchio commerciale
Corning 432129
Altezza
12.1 cm
Diametro sommità
24.1 cm
Colore
blue
Descrizione generale
Corning ice pans and buckets are multi-purpose pans and buckets that are a great non-toxic alternative to conventional ice pans. EVA has little or no odor, is inert and used in biomedical engineering applications such as drug delivery devices. The multi-purpose containers are highly insulative, stackable, unbreakable, lightweight and leak-proof. Containers will not "sweat" or deform and have a textured finish on the bottom to prevent slipping on the benchtop. Ideal for use with ice, dry ice (-78?), liquid nitrogen (-196?), alcohol or saline solutions, or warm solutions (+93°C). Available in five sizes and seven vibrant colors. It can be cleaned with soap and water, 10% bleach or 70% alcohol.
  • Non-toxic, recyclable ethyl-vinyl acetate (EVA) foam containers for use with ice, dry ice, liquid nitrogen, alcohol slurries.
  • Will not sweat, leak or skid on bench.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Global Trade Item Number
SKUGTIN
CLS432129-1EA4061835178858