Documenti fondamentali
CLS432076
Corning® CoolCell Filler Vials, for use with CoolCell LX and CoolCell FTS30 containers
2 mL
Sinonimo/i:
cryovials
About This Item
Codice UNSPSC:
41103025
NACRES:
NB.12
Confezionamento
pkg of 6
Produttore/marchio commerciale
Corning 432076
Capacità
2 mL
Descrizione generale
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. It is suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5ml LX containers.
Note legali
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
CLS432076-6EA4061835179312