Documenti fondamentali
CLS432075
Corning® ThermalTray
HP high profile for water bath
Sinonimo/i:
lab equipment accessory
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.12
Caratteristiche
HP high profile for water bath
Confezionamento
pkg of 1
Produttore/marchio commerciale
Corning 432075
Lungh. × Largh. × Alt.
28.0 cm × 14.0 cm × 8.6 cm
Descrizione generale
Corning ThermalTray thermo-conductive platforms support CoolRack and CoolSink sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
Note legali
Corning is a registered trademark of Corning, Inc.
ThermalTray is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)