Documenti
CLS431225
Corning® syringe filters
Nylon membrane, pore size 0.45 μm
Sinonimo/i:
disposable syringe filters, sterile syringe filters, syringe filters
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Materiali
Nylon membrane
polystyrene housing
Sterilità
sterile
Confezionamento
case of 50
Produttore/marchio commerciale
Corning 431225
Diametro
25 mm
Dimensione pori
0.45 μm
Raccordi
female Luer inlet (lock)
male Luer outlet (slip)
Descrizione generale
Corning syringe filters are 100% integrity tested and are certified nonpyrogenic and noncytotoxic and are manufacturered in accordance with ISO 9002 standards. A variety of membranes are available to meet your needs.
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
These Corning syringe filters have polypropylene copolymer housings, female luer lock inlets and male luer slip outlets.
  • Nylon (NY) membranes are naturally hydrophilic, surfactant-free and offer the lowest extractables
  • 100% integrity tested, sterile and certified nonpyrogenic and noncytotoxic
  • Manufactured in accordance with ISO 9002 standards
Note legali
Corning is a registered trademark of Corning, Inc.
Visita l’Archivio dei documenti
Nhan Huynh et al.
Nature communications, 10(1), 5463-5463 (2019-12-01)
Iron Regulatory Protein 1 (IRP1) is a bifunctional cytosolic iron sensor. When iron levels are normal, IRP1 harbours an iron-sulphur cluster (holo-IRP1), an enzyme with aconitase activity. When iron levels fall, IRP1 loses the cluster (apo-IRP1) and binds to iron-responsive