CLS3081250LO
Pyrex® large knob top desiccators
replacement desiccator bowl 10.5L large knob top desiccator, 1/cs
Sinonimo/i:
desiccator bowls, desiccator lower bowls
About This Item
Codice UNSPSC:
41104510
Prodotti consigliati
Materiali
borosilicate glass
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
Corning 3081-250LO
Compatibilità
suitable for (general analytical work)
Categorie correlate
Descrizione generale
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
This is the replacement desiccator bowl which fits Pyrex desiccator CLS3181250.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)