Documenti fondamentali
CLS2975223
Corning® cover glasses
rectangular, No. 1, W × L 22 mm × 30 mm
Sinonimo/i:
coverslips, glass covers, slide covers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.16
Materiali
borosilicate glass
rectangular
Confezionamento
pack of 100 ea
case of 1000 ea
Produttore/marchio commerciale
Corning 2975-223
Largh. × lungh.
22 mm × 30 mm
Tipo di binding
non-treated surface
Caratteristiche e vantaggi
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)