Documenti fondamentali
CAA444-EP
Caps for Eppendorf® A-4-44 Rotor
aerosol-tight, for 5804/5804R & 5810/5810R, for 100 mL rectangular buckets
About This Item
Codice UNSPSC:
41103913
Caratteristiche
aerosol-tight
for 100 mL rectangular buckets
Confezionamento
pack of 2
Produttore/marchio commerciale
Eppendorf® 022637428 (NA)
Eppendorf® 5804712005 (EU)
tecniche
centrifugation: suitable
Compatibilità
for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810
Categorie correlate
Note legali
Eppendorf is a registered trademark of Eppendorf AG