C9811
CryoCane™ aluminum canes
capacity, 5 vial(s)
Sinonimo/i:
cryogenic vial cane, cryostorage cane, cryovial cane, dewar cane, liquid nitrogen cane
About This Item
Codice UNSPSC:
41103029
NACRES:
NB.22
Prodotti consigliati
Caratteristiche
capacity 5 vial(s)
Produttore/marchio commerciale
Nalgene®
Lungh. complessiva
290 mm
Categorie correlate
Descrizione generale
Firmly holds 1.2 or 2.0 mL Nalgene cryogenic vials for storage in Dewar-type, liquid nitrogen freezers.
Projections support base of each vial.
Note legali
CryoCane is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)