Documenti fondamentali
BMSIPD960050S
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Hard Samples
stainless steel, capacity 50 mL, pack of 2
About This Item
Codice UNSPSC:
41100000
NACRES:
NA.82
Materiali
stainless steel
Confezionamento
pack of 2
Produttore/marchio commerciale
Benchmark Scientific IPD9600-50S
Capacità
50 mL
Compatibilità
BeadBlaster 96
Descrizione generale
Different materials have different grinding requirements. Very hard samples require a jar and grinding balls that are just as hard or harder for effective sample preparation. The jars feature threaded closures for leakproof performance with both solids and liquids. Stainless steel is a very hard material, and when combined with the stainless steel balls, provides for grinding of hard samples such as minerals, ores, polymers and circuit boards. Cryogrinding and also be performed with stainless steel jars and balls. These jars are available in 25ml and 50ml volumes with 10mm and 25mm grinding balls.
Applicazioni
Homogenization of cryo samples
Note legali
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)