AXYSCT050SSRS
Self standing screw cap tubes and attached caps with O-rings
capacity 0.5 mL, red, sterile
Sinonimo/i:
centrifuge tubes
About This Item
Materiali
red
screw top cap
Sterilità
sterile
Caratteristiche
RCF 20,000 × g
Confezionamento
pack of 500 tubes
case of 8 packs
Produttore/marchio commerciale
Axygen Inc
Corning
Capacità
0.5 mL