Documenti fondamentali
AXYP5ML48C100
Corning® Axygen® 48 Rectangular Well Deep Well Plate
well volume (5 mL), pyramid bottom, case of 100 ea
Sinonimo/i:
48-Well Deep Well Plate
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
pyramid bottom
Confezionamento
case of 100 ea
Produttore/marchio commerciale
Corning P-5ML-48-C-100
Volume pozzetto
(5 mL)
Descrizione generale
  • Axygen deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80 C.
  • <UL><LI>5.0 mL deep
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)