Documenti fondamentali
AXYAM750ULRD
Silicone sealing mat for minitubes, pierceable, 96 Elisa and storage plates, and 750 μL 96 well plates
pkg of 5x10mats/cs
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.22
Materiali
silicone (research-grade)
Sterilità
non-sterile
Produttore/marchio commerciale
Axygen Inc AM-750UL-RD
Corning
Confezionamento
pkg of 5x10mats/cs
Descrizione generale
  • Axygen® silicone sealing mat is ideal for use with 96 round well deep-well plates and 96-well ELISA plates.
  • Sealing mats reduce evaporation in PCR and storage applications
  • Made of autoclavable research-grade silicone
  • Reusable by following a bleach wash and ethanol rinse protocol.
  • Optimized to fit Axygen® MTS system, 1.0 mL Greiner and Corning deep well plates.
Note legali
Axygen is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)