VPMH105000
Viresolve® Pro Magnus Holder
Sinonimo/i:
Viresolve® Pro Magnus Holder (for 1 to 5 Devices)
About This Item
Codice UNSPSC:
23151806
eCl@ss:
32031690
Prodotti consigliati
Materiali
316L stainless steel component(s) (structural)
Livello qualitativo
Produttore/marchio commerciale
Viresolve®
Lungh. × Largh. × Alt.
135 cm (53 in.) × 61 cm (24 in.) × 56 cm (22 in.)
Descrizione generale
The Viresolve® Magnus Holders are designed for large volume processing. The Viresolve® Pro Magnus Holder is designed to run the Viresolve® Pro Device. No product contact eliminates the need for cleaning validations. This cartridge holder, VPMH105000, is capable of accepting 1 to 5 units
Caratteristiche e vantaggi
- Simple manual hydraulics to clamp units
- Vertical orientation to minimize footprint and facilitate use (stack, clamp, run)
Note legali
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany