Documenti fondamentali
NA18/25
Millipore
Diaphragm for NOVASEPTIC® Valve
PTFE
About This Item
Codice UNSPSC:
31411800
Materiali
PTFE
PTFE device (polytetrafluoroethylene)
Livello qualitativo
Caratteristiche
hydrophobic (Surface)
Parametri
-40-170 °C temp. range (Continous use)
-40-250 °C temp. range (Short term use)
Peso
0.02 kg (0.044 lb)
Applicazioni
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Confezionamento
The Diaphragm is packaged in a closed box.
Altre note
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Note legali
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany