MISELPRRI
MilliSentials Lab Printer Ribbon
About This Item
Codice UNSPSC:
44102408
NACRES:
IA.22
Prodotti consigliati
Diametro
× 1 in. , Ribbon Core (Inner Diameter)
Livello qualitativo
Dimensioni rotolo
1 , ∼6400 labels printed from 1 roll of ribbon
Descrizione generale
The MilliSentials Lab Printer Ribbon features a thermal transfer ink capable of withstanding extreme temperatures and harsh chemical solvents.
For use with the MilliSentials Lab Labeling System
For use with the MilliSentials Lab Labeling System
Applicazioni
Laboratory Sample Labeling