Z756369
Scienceware® Buchner funnel trowel
pkg of 1 ea
Sinonimo/i:
laboratory scoop
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F36820-0118
Lungh. × Largh. × Alt.
19 cm (7 1/2 in.) × 7.6 cm (3 in.) × 12 cm (4 3/4 in.)
Peso
0.8 lb
Categorie correlate
Descrizione generale
The Scienceware® Buchner Funnel Trowel is specifically designed for use with 18, 24 and 36 in. Buchner Funnels. Just apply slight downward pressure and ′sweep′ the trowel along the surface of the precipitate to quickly and efficiently smooth fissures appearing during the filtration process.
- Large surface area smooths fissures faster than traditional narrow spatula method
- Dual-beveled edges for bi-directional smoothing
- Curved ends match Buchner Funnel sidewalls allowing easy access to entire cake surface
- FDA-grade high-density polyethylene (HDPE) complies with industry standards and provides maximum chemical-resistance
- Welded construction with no hollow cavities assures hygienic use and easy cleaning
- Compatible with industrial dishwasher temperatures and cleaners
- Trowel base: 1.3 cm H (1/2 in.)
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z756369-1EA | 4061832266978 |