Z744039
KitAlysis Vials
Sinonimo/i:
Pack of 96 shell vials (8x30) with stackable tray
Descrizione generale
KitAlysis Vials is a pack of 96 shell vials that are compatible with the KitAlysis High-Throughput Screening Platform. These vials allow the user to screen their own catalysts and ligands using the KitAlysis 24-well Reaction Block.
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)