Z743179
Julabo® bath covers
lift-up gable cover to fit CORIO CD-B19/27 and CD-BT19/27
About This Item
Codice UNSPSC:
41103706
NACRES:
NB.77
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Julabo 9970254
Descrizione generale
The hinged stainless steel gable hood reduces the evaporation of tempering liquid and saves energy. The rising steam is led back into the bath via a drip edge. This means that the tempering liquid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath. The hinged stainless steel gable hood is designed for operation up to +60°C.
Note legali
Julabo is a registered trademark of Julabo USA, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)