Key Documents
Z742869
Kern PLS series precision balances
Kern PLS 1200-3A, weighing capacity 1200 g, accuracy: 0.001 g, precision: ±0.003 g
Sinonimo/i:
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
Codice UNSPSC:
41111502
NACRES:
NB.71
Ingresso CA/CC
110 - 240 V AC, universal plug set
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Kern PLS 1200-3A
Capacità di pesata
1200 g, accuracy: 0.001 g, precision: ±0.003 g
Descrizione generale
Range of precision balances with CAL adjusting program for quick setting of the balance accuracy.
Ergonomically optimised keypad for left and righthanded users
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)