Documenti fondamentali
Z741011
Sigma-Aldrich
Aldrich® Essentials round media storage bottle
wide mouth, capacity 100 mL, GL 45 polypropylene cap with pour ring
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
borosilicate glass bottle
polypropylene cap
round bottle
Giunto
Joints threaded neck (GL45)
Caratteristiche
graduations
with cap
Confezionamento
pkg of 10 ea
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
100 mL
Descrizione generale
Borosilicate glass, wide mouth, media/reagent bottles with polypropylene GL 45 thread screw cap with pour ring. Large marking spot and reference graduations.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)