Z684457
Aldrich® solvent storage/dispensing flask, septum-inlet, with PTFE inlet valve
joint: ST/NS 29/32, capacity 100 mL
Sinonimo/i:
solvent dispenser bottles
About This Item
Prodotti consigliati
Materiali
glass
round bottom flask
single-neck
Giunto
joint: ST/NS29/32
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
case of 1
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
100 mL
Descrizione generale
Designed for use with PureSolv Micro systems with a standard taper dispensing joint. PTFE inlet valve may be closed after filling to isolate dry solvent and to remove flask from system and take to another location for use. Use septum Z565695 or Z565679.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)