Documenti fondamentali
Z648442
Quickfit® bends with vacuum connection
socket joint: ST/NS 19/26, cone joint: ST/NS 24/29
Sinonimo/i:
RA3/23/SC
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
cone joint: ST/NS24/29
socket joint: ST/NS19/26
Produttore/marchio commerciale
Quickfit RA 3/23/SC
Disponibilità
not available in USA and Canada
not available in USA and Canada
Descrizione generale
Socket to cone. With screw-thread adapter for flexible tubing.
Caratteristiche e vantaggi
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Items marked with * complies with DIN 12594• Designed to fit the angled condenser in vacuum distillation applications• Delivery stem within cone• Supplied with detachable plastic screw thread connectors which allow 9mm bore flexible tubing to be fitted easily and safely
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)