Key Documents
Z647543
Pyrex® measuring cylinder with spout and detachable plastic foot, complete
volume 250 mL
Sinonimo/i:
3205/12P
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Caratteristiche
graduations 2.0 mL
Campo di misura del volume
250 mL
Altezza
320 mm
Descrizione generale
  • Complies with ISO 4788
  • Glass cylinder has blue enamel graduations
  • Polyethylene foot absorbs impact and reduces breakage
Note legali
PYREX is a registered trademark of Corning, Inc.