Z557587
Aldrich® pear-shaped splash-guard adapter
capacity 100 mL, female joint: ST/NS 24/40, male joint: ST/NS 24/40
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
female joint: ST/NS24/40
male joint: ST/NS24/40
Descrizione
anti-climb
Capacità
100 mL
Descrizione generale
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)