Z553387
Aldrich® automatic shut-off quick-disconnect
Coupling body
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Descrizione
1/4in. I.D. tubing connection
Descrizione generale
Replace SafetyBarb tubing connectors with these new quick-disconnect fittings, which are made specifically for use with Aldrich glassware. To install quick-disconnects, unscrew tubing connectors on glassware and replace with coupling inserts Z55,337-9. Order one coupling insert and one coupling body for each glass connection. Chemically resistant acetal coupling insert and body are spring-loaded, locking, and have 316SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically. For use with flexible 1/4 in. I.D. tubing.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z553387-1EA | 4061832965178 |