Z548820
Aldrich® solvent-pouring funnel
stem L × O.D. 100 mm × 22 mm
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Lungh. stelo × D.E.
100 mm × 22 mm
Categorie correlate
Descrizione generale
Unique, closed-end glass funnel has 2 mm diameter holes in the stem permitting the gentle addition of solvents to chromatography columns without disturbing the silica gel/sand layer. The top of the funnel is 75 mm in diameter.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)