Documenti fondamentali
Z530719
Sigma-Aldrich
Aldrich® vacuum manifold with joints
positions, 5, joint: ST/NS 24/40
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.79
Giunto
joint: ST/NS24/40
Caratteristiche
positions 5
Lunghezza
500 mm
Descrizione generale
This practical design provides clearance to accommodate 250mL flasks with a snap of a Keck® clip. Manifolds are available with either <ST/>14/20, <ST/>24/40, or <ST/>29/32 joints. Glass stopcocks have a 4mm bore.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z530719-1EA4061832132662