Z512966
Aldrich® corrosive lecture-bottle gas regulator
delivery pressure 0-15 psig, CGA Inlet size 170
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Prodotti consigliati
delivery pressure measuring range
0-15 psig
Dimens. Ingr. CGA
170
Descrizione generale
- Single-stage
- CGA inlet connection, needle valve outlet with ¼ in. NPTM connection
- Max. inlet pressure: 0-3000 psig
- Temp. range: -40 to 140°F
- 316 Stainless Steel body 1 ½ in. gauge face, PTFE seals and inner friction sleeves
- NPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure; order bonnet-vent tubing connectors separately
- Use with "T"-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipment
- Use for high-purity applications
Lecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.
Altre note
See Technical Information Bulletin AL-151 Gas Regulators: Selection, Installation and Operation.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z512966-1EA | 4061833351758 |