Key Documents
Z504858
Sigma-Aldrich
Ace O-rings, FETFE®
I.D. 9.2 mm, wall size 2.6 mm
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Confezionamento
pkg of 12 ea
Produttore/marchio commerciale
Ace Glass 7855716
Diam. int.
9.2 mm
Misura parete
2.6 mm
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Ace pressure tube bushing type, Front seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Z181102
Ace pressure tube
Ace O-rings, FETFE® I.D. 12.4 mm, wall size 1.78 mm
Z504742
Ace O-rings, FETFE®
Ace pressure tube bushing type, Back seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Z181072
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~38 mL, L × O.D. 20.3 cm × 25.4 mm
Z181110
Ace pressure tube
Ace pressure tube Front seal, capacity 120 mL, 150 PSI, #15 plug, L × O.D. 17.8 cm × 38.1 mm
Z566268
Ace pressure tube
Ace pressure tube bushing type, Back seal, capacity ~38 mL, L × O.D. 20.3 cm × 25.4 mm
Z181080
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~9 mL, L × O.D. 20.3 cm × 13 mm
Z564613
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity 60 mL, L × O.D. 10.2 cm × 38.1 mm
Z568767
Ace pressure tube