Key Documents
Z414255
Sigma-Aldrich
Aldrich® septum-inlet adapter, in-line
Plain, Joints: ST/NS 24/40 (2)
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
Joints: ST/NS24/40 (2)
Categorie correlate
Descrizione generale
Inlet diam. 14 mm. Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)