Documenti fondamentali
Z341886
IKA® fixing clip attachment for KS 130 basic and control shakers
AS 130.2 fixing clip attachment
Sinonimo/i:
flask clamps, holding clamps
About This Item
Codice UNSPSC:
41103811
NACRES:
NB.73
Confezionamento
set of 1
Produttore/marchio commerciale
IKA 3115000
Descrizione generale
Note: This attachment (without fixing clips) is for processing round flasks, measuring flasks and Erlenmeyer flasks.
Note legali
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)