Documenti fondamentali
Z303399
Quickfit® double surface (Davies) condenser
jacket L 205 mm, joint: ST/NS 29/32
Sinonimo/i:
C5/24/SC
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Giunto
joint: ST/NS29/32
Produttore/marchio commerciale
Quickfit C 5/24/SC
Disponibilità
not available in USA and Canada
not available in USA and Canada
Lunghezza camicia
205 mm
Area superficiale
300 cm2
Descrizione generale
Increased surface area and effectiveness when compared with the corresponding Liebig condenser. Inner cooling surface gives a baffling effect, while outer cooling surface prevents creep and loss of vapour when used with low boiling liquids.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)