Documenti fondamentali
Z303011
Quickfit® pear-shaped flasks
capacity 100 mL, joint: ST/NS 14/23
Sinonimo/i:
FP100/1
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
pear-shaped flask
single-neck
Giunto
joint: ST/NS14/23
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
pack of 1
Produttore/marchio commerciale
Quickfit FP100/1
Disponibilità
not available in USA and Canada
Capacità
100 mL
Descrizione generale
Single neck with ground-glass joint
Caratteristiche e vantaggi
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Ideal for micro and semi-micro distillations.• Reduced losses from thermal decomposition• Minimum hold-up
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)